United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
11 >
Representative photo only
Cole Parmer Sealless Centrifugal Pump Self Priming PVDF Magnetic Drive 110GPM 72FT three phase 2HP TEFC Motor
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-72225-30
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Centrifugal Pumps
>
Centrifugal Pumps that are Magnetically Coupled (have a Magnetic Seal)
>
Cole-Parmer Sealless Centrifugal Pumps, Self-Priming PVDF Magnetic Drive
Share
|
Cole-Parmer Sealless Centrifugal Pump, Self-Priming PVDF Magnetic Drive, 110GPM - 72FT, three phase, 2HP, TEFC Motor
EW-72225-30
$4,294.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Experience up to 70% operating efficiency in addition to greater reliability compared to sealed pumps
Powerful, rare earth magnetic coupling uses standard NEMA (60 Hz) motor
Greater efficiency translates into reduction in energy usage
High working pressure safely handles high specific gravity fluids
Superior lift capability lifts fluid from as deep as 25 feet below the pump
Fast primingprimes from 18 feet in as little as 90 seconds
These Cole-Parmer horizontal, self-priming, PVDF magnetic drive centrifugal pumps offer many features which improve performance. The volute and impeller design allow for high efficiency, resulting in energy usage reduction, especially when compared to other established pump technologies. As a result of the high working pressure, high specific gravity fluids can be safely pumped. Another benefit is the protection against unexpected surges in system pressure. The standard NPT or BSP connections allow for easy installation. The wetted materials of construction were chosen and engineered for aggressive liquids.
Pumps feature carbon bushings that allow extended run-dry ability. The thrust balance system has generous balance holes and rear sealing ring to reduce axial thrust. The replaceable dual bushings ride evenly on the shaft for optimal alignment and reliability. The internal spiral groove helps to flush and lubricate the bushings during operation. PTFE bushings are the ideal choice for chemicals like sodium hypochlorite (bleach), chromic acid, chrome plating solutions, and nitric, chromic, and hydrofluoric acids.
Self-priming centrifugal pumps are extremely versatile. The priming ability allows the pumps to handle applications difficult for standard centrifugal pumps, such as pumping fluid from below grade, emptying tanker trucks or rail cars, and transferring fluids from the top of tall bulk storage tanks to reduce the possibility of leaks from sidewall-mounted bulkhead fittings.
What's Included:
Conduit box for hardwiring.
Specifications
Product Type
Magnetic Drive Centrifugal Pumps
Max flow rate (GPM)
110
Max flow rate (LPM)
416
Max head (ft)
72
Max head (m)
21.9
Max system pressure (psi)
90
Motor (phase)
3
Motor (rpm)
3450
Motor (VAC)
230/460
Motor (Hz)
60
Motor (Amps)
5.15/2.58
Motor (hp)
2
Motor (type)
TEFC
Wetted materials
Carbon-filled PVDF impeller, 99.9% alumina ceramic, PTFE bushing, Viton® O-ring
Max fluid temp (º F)
220
Max fluid temp (º C)
104
Duty cycle
Continuous
Connections (inlet)
1-1/2" NPT(F)
Connections (outlet)
1-1/2" NPT(M)
Suction lift
25 ft H
2
O
CE Compliance
Yes
Dimensions
22-25/32"L x 9-23/32"W x 11-1/8"H
(57.8 x 24.7 x 28.2 cm)
Brand
Cole-Parmer
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Centrifugal Pump Selection Guide
Gravity Feed Installation of Your Centrifugal Pump
How to Read a Pump Curve
Show All...
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title