dummy
EW-59986-01
- Quiet, electromagnetic vibratory action for small samples
- 0 to 100% amplitude control for fine particle separation
This sieve shaker accomodates up to seven full-height 3''-diameter sieves and is ideal for small samples of chemicals, minerals, pharmaceuticals, powered metals, cosmetics, abrasives, ores, foods and other fine powders. The stainless steel, 3''-diameter sieves, sieve cover, and sieve pan are sold separately.
High frequency, electromagnetic vibratory action with 0 to 100% amplitude control is ideal for fine particle separations. Inline solenoid-actuated tapping action may be used during vibration to speed dry separations. Also use the tap separately for tap-settling and bulk density tests.
The unique clamping mechanism makes insertion and removal of the sieves simple and convenient and holds the sieves securely during operation. Shaker also features on/off controls for vibration and a manual timed-mode switch. Digital timer shows time set and time remaining to enable reliable, repeatable results.
What's Included:
6-ft power cord. Sieves sold separately.
| Product Type |
Sieve Shakers |
| Sieves size accepted (" OD) |
3 |
| Dimensions (" W) |
8 |
| Dimensions (" H) |
22 |
| Dimensions (" D) |
11 |
| Dimensions (cm) |
20.3 W x 55.9 H x 27.9 D |
| Power (Hz) |
50/60 Hz |
| Timer range (min) |
1 second - 99 minutes plus .03% repeat accuracy |
| Power (VAC) |
115 |
| Vibrational frequency |
3600 vpm |
| Frequency |
60 taps/min |
| Shipping weight (lbs) |
16 |
| Shipping weight (kg) |
7.3 |
| Brand |
Cole-Parmer |
| Manufacturer number |
SS-3 |
at $52.00 USD/EACH
Usually ships in 15 days.
at $52.00 USD/EACH
Usually ships in 14 days.
at $52.00 USD/EACH
Usually ships in 14 days.
at $52.00 USD/EACH
Usually ships in 13 days.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
Usually ships in 14 days.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $52.00 USD/EACH
In stock.
at $56.00 USD/EACH
In stock.
at $56.00 USD/EACH
In stock.
at $56.00 USD/EACH
Call 1-888-358-4705 for availability.
at $62.00 USD/EACH
In stock.
at $62.00 USD/EACH
In stock.
at $63.00 USD/EACH
In stock.
at $63.00 USD/EACH
In stock.
at $63.00 USD/EACH
In stock.
at $82.00 USD/EACH
In stock.
at $22.00 USD/EACH
In stock.
at $48.00 USD/EACH
In stock.
at $1,358.30 USD/EACH
In stock.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection