Cole-Parmer Solenoid Pinch Valve; for use with L/S 25 tubing, normally closed, 12 VDC
EW-98306-16
$138.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Qty
  • Two-way pinch valves specifically designed to work with Masterflex® pump tubing
  • Use with tubing that has durometer rating of 50 or less such as C-Flex® or silicone—7.25 psi max
  • Excellent for clean processes—fluid stays contained within tubing
  • Use for on/off flow control applications
Tube size L/S® 25
Watts 10
Response 30 to 50 msec
Power (VDC) 12
Brand Cole-Parmer
Model PK-4805NC-12V
Product Type Solenoid Electric Valves (actuated)
Manufacturer number PK-4805NC-12V
Related products
Qty/Price
Availability
at $45.50 USD/EACH
In stock.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  • 13-14 Spanish Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection