Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Cole-Parmer Solid Black Rubber Stoppers, Standard Size 2; 52/Pk

Item# EW-62990-08

$40.25 USD/PACKAGE of 52

Quantity Discounts
Qty: 6 30 120+
Price: $36.22 $34.21 $32.20
    Black rubber stoppers stay pliant over a long period and withstand laboratory conditions without hardening. Stoppers retain resiliency even after repeated autoclaving. Each stopper has its size in raised figures on top. Contain some sulfur. Autoclavable.
    Product Type Stoppers
    Standard size number 2
    Top OD (mm) 20
    Color Black
    Material Rubber
    Bottom OD (mm) 16
    Qty/pk 52
    Brand Cole-Parmer

    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought