United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 9
of
10 >
SI-27018-40
Cole-Parmer
®
Solution Grounded, Platinum-Band ORP Electrode; No ATC Element, BNC
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-27018-40
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Electrodes, Industrial
>
pH/Ion Industrial Electrodes
>
Cole-Parmer In-line or Submersible, Solution Grounded pH/ORP Electrodes
Share
|
Cole-Parmer Solution Grounded, Platinum-Band ORP Electrode; No ATC Element, BNC
SI-27018-40
$241.00 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
6
10%
$216.90/EACH
11
15%
$204.85/EACH
25
20%
$192.80/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton
®
housing with 3/4'' NPT(M) threads.
General-Purpose Electrodes
have low-resistant glass with a very fast and stable response. Designed for minimal salt applications. Use for measurements from 0 to 13 pH. Temperature range is 32 to 212°F (0 to 100°C).
High-Salt Application Electrodes
are designed for continuous long-term use at high temperatures in strong alkaline solutions above pH 11. Use for measurements from 0 to 14 pH. Negligible salt ion error may be expected above pH 13. Response time will increase as solution temperature rises due to high glass impedance. Temperature range is 32 to 275°F (0 to 135°C).
Platinum/Gold-Ring ORP Electrodes
have a metal-band sensor for oxidation-reduction measurements. Temperature range is 32 to 266°F (0 to 130°C).
Specifications
ATC element
None
Temp range
32 to 266°F (0 to 130°C)
Junction
Double
Connector
BNC
Applications
Oxidation-reduction measurements
Manufacturer number
H721S90003T10AB
Brand
Cole-Parmer
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Tech Challenge: Faulty pH Readings
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title