Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Cole-Parmer StableTemp Digital Utility Water Baths, 2 liters, 120V, 60 Hz
Item# EW-14576-00
$389.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 10 days or sooner.
Qty
  • Offering digital technology at analog prices
  • Easy-to-read LCD display shows your choice of °C or °F
  • Temperature can be recalled at the touch of a button — program up to 3 commonly used setpoints
  • Built-in timer notifies you when your process is complete
  • High-temperature alarm and safety shutdown features keep you and your bath safe and working
StableTemp utility baths combine precise temperature control and excellent temperature stability to provide you with reliable heating. These baths feature dual thermostats — a primary microprocessor-based temperature controller that provides ±0.1°C precision, and a secondary over-temperature safety sensor prevents samples from overheating. For optimal temperature accuracy, the primary controller can be calibrated to your reference thermometer.
The stainless steel tank is seamless — no welds to leak or rust — and the rounded corners provide an easy-to-clean interior. Hinged polycarbonate lid allows condensation to flow back.
What's Included:
gabled polycarbonate cover, sample shelf, and 6-ft (1.8-m) power cord.
Product Type Water Bath
Bath type Utility Water Bath
Capacity (Liters) 2
Max temperature (º C) 100
Dimensions (" W) 11.5
Dimensions (" H) 12
Dimensions (" L) 9
Min temperature (º C) Ambient +5°
Temp control PID
Temp display LED, switchable between C and F
Temp sensor 100 ohm PT RTD
Temp stability ±0.1°C
Uniformity ±0.2°C
High-temperature cutoff Yes
Working area (" W) 4.3
Working area (" L) 3.9
Working area (" D) 5.5
Qty/ea 1
Wetted materials 304 Stainless Steel
Brand Cole-Parmer StableTemp
Heater wattage 120
Power (VAC) 120
Power (Hz) 60
CE Compliance Yes
Accessories
Qty/Price
Availability
at $1,035.00 USD/GALLON
Available for order
or call 1-888-358-4705 to order
at $591.00 USD/GALLON
Available for order
or call 1-888-358-4705 to order
at $451.00 USD/GALLON
In stock.
at $210.00 USD/GALLON
In stock.
at $247.00 USD/PACKAGE
In stock.
at $110.00 USD/PACKAGE
In stock.
at $157.00 USD/PACKAGE
In stock.
at $279.00 USD/PACKAGE
Usually ships in 8 days.
at $454.00 USD/PACKAGE
Usually ships in 8 days.
at $43.00 USD/EACH
In stock.
at $28.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability
at $266.00 USD/EACH
Usually ships in 10 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
2 Questions | 2 Answers
Displaying questions 1-2
  • Q:
    Min temperature "Ambient +5C".
    Does it mean it can cool down to +5C when room temperature is +20C?
    Asked on 4/29/2016 by Natalya from United States
    1 answer
    • STAFF EXPERT
      A:
      No, this means that the temperature range at the low end will be 5C above ambient (room temperature), hence ambient + 5C.
      Answered on 4/29/2016 by Rishi from Technical Support
  • Q:
    What are the internal dimensions of the water bath?
    Asked on 4/29/2015 by SSS from CA
    1 answer
    • STAFF EXPERT
      A:
      The internal reservoir dimensions are 3.9 x 4.3 x 6" (9.9 x 10.9 x 15.2 cm)
      Answered on 4/29/2015 by Anonymous
Displaying questions 1-2
Do you have a question about this product? 