United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Cole Parmer StableTemp Dry Block Heaters with Heated Lid Digital 2 block 120V
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36600-69
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Block Heaters
>
Cole-Parmer StableTemp Dry Block Heaters and Blocks
>
Cole-Parmer StableTemp Dry Block Heaters with Heated Cover (Digital)
Share
|
Cole-Parmer StableTemp Dry Block Heaters with Heated Lid (Digital), 2 block, 120V
EW-36600-69
$960.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Precision temperature stability ensures reproducibility of results
Create a versatile system unique to your application; order a digital heater and the required heating block module (sold separately online).
different-size tubes -order blocks separately
Designed for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a wide variety of other laboratory procedures. Each heater accepts separate interchangeable modular heating blocks, which accommodate various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes. Modular blocks are constructed from a solid anodized aluminum block. The close tube to block walls allow for maximum heat retention. Each block has a thermometer well for measuring block temperature. Optional PT100 RTD temperature probe kit enables the unit to read the actual block or sample temperature and display the temperature on the control panel. Create a versatile system unique to your application by selecting the modular blocks for your application.(blocks sold separately online).
CAUTION! To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed to run as a dry bath / incubator.
Digital Dry Block Heaters offer exceptional temperature uniformity and feature a proportional integrative derivative (PID) controller. PID control allows you to select a particular temperature with ease (in 0.1°C increments), enables rapid heat-up for samples, and provides excellent stability to maintain results. Features include timer, audible alarm, built-in temperature calibration mode, overshoot protection, and independent LED displays that show actual and setpoint temperatures
Digital dry block heater with heated lid, 2 block capacity 120V with independent LED displays for temperature & time.PID temperature control with user calibration mode.
Specifications
Product Type
Dry Block Heaters-Digital
CE Compliance
Yes
Uniformity
± 0.2°C
Watts
400
Max temperature (º F)
212
Power (VAC)
120
Max temperature (º C)
100
Dimensions
15.4Lx8Wx7H
Number of Blocks
2
Brand
Cole-Parmer
Manufacturer number
949302HLCP
Model
949302HLCP
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title