United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
10 >
Representative photo only
Cole Parmer StableTemp Economy bath 2L 120V 60Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-12106-70
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Laboratory, Utility Water Baths
>
Cole-Parmer StableTemp Economical Water Baths
Share
|
Cole-Parmer StableTemp Economy bath, 2L, 120V, 60Hz
EW-12106-70
$589.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Our best selling utility baths offer excellent performance at affordable prices!
±0.2°C stability at 37°C
Seamless 304 stainless steel tank and powder-coated exterior are corrosion resistant
Front-panel controls are safe and easy-to-use
These water baths are ideal for general-purpose applications. Bath fluids can be controlled at temperatures as high as 100°C (60° without cover) with +/-0.2° precision and uniformity. Bath temperature can be read from alcohol thermometer (included) with a range from -20 to 150°C. The steeply gabled, polycarbonate cover accommodates glassware of varying heights and tilts out of your way when loading or removing samples, allowing condensate to drain neatly back into the bath.Laboratory, utility water baths are the ideal bath for maintaining a temperature set points over a long period of time. Different size and depth of the bath chamber to meet most every application. Units feature analog or digital temperature control with a ranges from ambient to 100° C. Options of high-temperature and low-level cutoff features to protect samples.
What's Included:
cover, glass thermometer, and a 3-ft (0.9-m) power cord with three-prong plug (European plug on 240 VAC models.
Specifications
Product Type
Water Bath
Bath type
Utility Water Bath
Capacity (Liters)
2
Min temperature (º C)
Ambient
Max temperature (º C)
100
Temp control
Stable on/off control
Temp sensor
Thermistor
Temp stability
(at 37°C) ±0.2°C
High-temperature cutoff
Yes
Temp setting
Analog
Bath opening
5.31 x 5.88 x 5.81”
Working area (" D)
5.81”
Wetted materials
304 SS
Power (VAC)
120V
Power (Hz)
60Hz
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Keeping Samples the Right Temperature
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Show All...
Bath and Circulator Specifications Terminology
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title