United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
7 >
Representative photo only
Cole Parmer StableTemp Stainless Steel Sand Baths 1 block models
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36600-57
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Block Heaters
>
Cole-Parmer StableTemp Dry Block Heaters and Blocks
>
Cole-Parmer StableTemp Dry Block Heater, Modular Heating Blocks
>
Cole-Parmer StableTemp Dry Block Heater, Stainless Steel Sand Baths Heating Blocks and Accessories
Share
|
Cole-Parmer StableTemp Stainless Steel Sand Baths, 1 block models
EW-36600-57
$226.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Stainless Steel Sand Baths are ideal for irregular vessels or to hold a variety of different size vessels at one time. Constructed of stainless steel for superior corrosion resistance. Designed to hold sand, stainless steel shot, or non-volatile liquids. Fits into the Cole Parmer Stable Temp Dry Block Heaters in place of a modular block. Select the appropriate capacity for the size block heater.Stainless Steel Sand Baths are ideal for irregular vessels or to hold a variety of different size vessels at one time. Constructed of stainless steel for superior corrosion resistance. Designed to hold sand, stainless steel shot, or non-volatile liquids. Fits into the Cole Parmer Stable Temp Dry Block Heater one block capacity models in place of a modular block.
Specifications
Product Type
Sand Bath Heating Block
Dimensions
3.75Lx3Wx2.5H"
Brand
Cole-Parmer
Manufacturer number
949132
Model
949132
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title