United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 9
of
12 >
Representative photo only
Cole Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer 3 1 2 gal 115 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-08895-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Ultrasonic Cleaners
>
Standard Ultrasonic Cleaners
>
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer
Share
|
Cole-Parmer Stainless Steel Ultrasonic Cleaner with Heater and Mechanical Timer, 3 1/2 gal, 115 VAC
EW-08895-50
$1,613.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Aggressive cleaning ability in a variety of sizes
Sturdy, noncorrosive units handle all types of cleaning tasks
Sweep frequency ensures uniform cleaning activity
Operating at 40 kHz, these ultrasonic cleaners provide aggressive cleaning power. Easy-to-clean, corrosion-resistant 304 stainless steel tanks have no corner welds or other dirt-catching surfaces. The units are enclosed in durable, powder-coated steel skirts.
Cleaners with heater and mechanical timer
offer greater cleaning capabilityideal for heat cleaning any size parts. Maximum heater temperature is 140°F (60°C). Set the cleaning cycle up to 60 minutes or use the hold function for continuous operation.
What's included:
stainless steel lid, nonskid feet, and a 7-1⁄2 ft power cord. The 1, 1-1⁄2, 2-1⁄2, 3-1⁄2, and 5-1⁄2 gallon cleaners have a bottom-mounted drain and include tank drain kits.
The 1-1/2-, 2-1/2-, and 5-1/2- gallon cleaners have a quick-disconnect drain.
The 1/2- and 3/4-gallon cleaners have a pour spout.
Never use ultrasonic cleaners with flammable solvents. A cleaning solution must be used with these cleaners. Use an insert tray or beaker positioning cover and beaker with these ultrasonic cleaners. Ultrasonic acticity is greatest starting about an inch from the bottom of the tank.
Specifications
Product Type
Ultrasonic Cleaner
Capacity (gal.)
3.5
Tank dimensions (" L)
11.5
Tank dimensions (" W)
9.5
Tank dimensions (" H)
8
Frequency output (Hz)
40000
Power (Amps)
4.2
Power (VAC)
115
Brand
Cole-Parmer
Manufacturer number
CPN-956-651
Accessories
Qty
/
Price
Availability
Solid stainless steel pan for Cole-Parmer 2 1/2 Gallon and Stainless Steel 2 1/2 & 3 1/2 Gallon Cleaners.
EW-08853-41
at
$133.00 USD/EACH
In stock.
Add to Cart
Perforated tray for Cole-Parmer 2 1/2 Gallon and Stainless Steel 2 1/2 & 3 1/2 Gallon Cleaners.
EW-08853-42
at
$149.00 USD/EACH
In stock.
Add to Cart
Beaker positioning cover for Cole-Parmer 2 1/2 Gallon and Stainless Steel 2 1/2 & 3 1/2 Gallon Cleaners, holds four 600-mL beakers
EW-08893-50
at
$61.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Optimizing your Ultrasonic Cleaner
Ultrasonic Cleaner FAQs
Ultrasonic Cleaner Maintenance
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title