Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Cole-Parmer Standard Benchtop Chilling/Heating Block, 100-230 VAC

EW-44175-00
$1,089.00 USD/EACH

Usually ships in 7 days.
Qty

  • Chill, freeze, heat, or mix biological samples at your work station
  • Store 5 programs in memory for precisely timed and repeatable routines
  • Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
  • Two-block unit offers two different or same temperatures
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.
Standard Chilling/Heating Block is ideal for a variety of applications such as storing enzymes or DNA libraries, maintaining 14°C for ligation reactions, and incubating or deactivating enzyme reactions.
What's Included:
Chiller/heater module, power cord, instruction manual, and a 12 VDC power supply. Order cover and aluminum blocks separately.
Product Type Chilling/Heating Blocks
Description Standard Chilling/Heating Block
Dimensions (overall) 8-3/4" L x 6-1/2" W x 3-1/2" H (22.2 x 16.5 x 8.9 cm)
Power (VAC) 100 to 230
Stability ±1°C
Temp display Two line LCD
Temp range -10 to 100°C
CE Compliance Yes
Brand Cole-Parmer
Manufacturer number IC20CP

Accessories
Qty/Price
Availability

at $161.00 USD/EACH

Usually ships in 15 days.

at $176.00 USD/EACH

In stock.

at $231.00 USD/EACH

Usually ships in 12 days.

at $221.00 USD/EACH

Call 1-888-358-4705 for availability.

at $186.00 USD/EACH

Usually ships in 9 days.

at $151.00 USD/EACH

Usually ships in 8 days.

at $156.00 USD/EACH

Usually ships in 7 days.

at $55.00 USD/EACH

Usually ships in 9 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title