United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
8 >
EW-04803-01
04803-05 shown in image
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04803-01
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17002-27
Innocal NIST Calibration for Stirrers and Heat
$160.00 USD/EACH/EACH
Usually ships in 16 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Stirrers
>
Stirring Hot Plates
>
Cole-Parmer Magnetic Stirring Hot Plate
Share
|
Cole-Parmer Stirring Hot Plate, 6" x 6", Aluminum top, 115 VAC, 60 Hz
EW-04803-01
$390.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Stepless speed control provides smooth stirring action to 1500 rpm
Unique U-shaped magnet holds stir bar tightly
Use our economical stirrer for all of your magnetic stirring applications—from gentle agitating to vigorous churning. The large, permanent drive magnet ensures positive coupling of drive to stir bar. Permanently lubricated, brushless drive motor provides long, trouble-free operation. Stirring hot plates are available wtih anodized aluminum or ceramic top plates. Choose the corrosion-resistant ceramic for your higher temperature applications.
Specifications
Product Type
Stirrer (Hot Plates)
Max load capacity (lbs)
15
Speed (rpm) (high)
1500
Max temperature (º C)
371
Top plate material
Aluminum
Top plate dimensions (in.)
6 x 6
Dimensions
7"W x 4.5"H x 10"D
Max stirring volume (Liters)
2.5
No. of stirring positions
1
Brand
Cole-Parmer
Manufacturer number
HS1A
Power
115VAC; 60Hz;
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Stirrers and Heat
EW-17002-27
at $160.00 USD/EACH
Usually ships in 16 days.
Add to Cart
Technical Resources
Instruction Manual.pdf
Cole-Parmer
Stirrers and Stirring Hot Plates (375 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title