United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
8 >
EW-10000-53
Cole-Parmer
®
Symmetry
®
IS Compact Industrial Bench Scale, 15 Kg X 0.5 G 115V. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-10000-53
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Industrial Compact Scales
>
Cole-Parmer Symmetry Industrial Compact Sales
>
Cole-Parmer Symmetry IS Compact Industrial Bench Scales
Share
|
Cole-Parmer Symmetry IS Compact Industrial Bench Scale, 15 Kg X 0.5 G 115V
EW-10000-53
$250.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Basic weighing and simple check weighing in a compact design
Large backlit display for quick reading
Built-in High/Low/OK indicators for fast over/under weighing
Display hold function freezes display when load is removed
Large stainless steel pan
These affordable compact bench scales handle basic weighing tasks and check weighing operations with a simple keypad and intuitive operation. They offer four weighing units, % weighing, simple parts counting, and check weighing.
What's Included:
AC power adapter and internal rechargeable battery
Specifications
Product Type
Compact Industrial Scales
Calibration
External
Capacity (kg)
15
Capacity (lbs)
33.1
Housing dimensions
25.4 x 10.8 x 30.5 cm
Dimensions (" W)
10
Dimensions (" H)
4-1/4
Dimensions (" D)
12
Display
LCD
Platform size (L x W)
10" x 7" (25 x 18 cm)
Readability (g)
0.5
Linearity (g)
±4
Repeatability (g)
±0.5
Weighing units
g, kg, lb, oz, lbs: ozs
Weighing modes
weighing, parts counting, percent weighing
Power (VAC)
120
Battery
internal rechargeable battery
Brand
Symmetry
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title