United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
SI-10000-16
Cole-Parmer
®
Symmetry
®
PA-Analytical Balance, 220g x 0.0001g -- 115 VAC. Representative Photo Only
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-10000-16
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Analytical Balances
>
Cole-Parmer Symmetry Analytical Balances
>
Cole-Parmer Symmetry PA Analytical Balances
Share
|
Cole-Parmer Symmetry PA-Analytical Balance, 220g x 0.0001g -- 115 VAC
SI-10000-16
$1,820.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Affordable analytical balances for precise weighing of small samples
All glass 3-door draft shield
Internal automatic calibration
Fast stabilization and response time
RS-232 Interface
Parts counting, percent weighing
Weighing units- g, ct, mg, dwt, grains, oz, troy ounces
Cole-Parmer’s precision analytical Symmetry balances are built with the most advanced weighing technology available today, offering durability, and easy-to-use operation. These balances feature fast reponse times—stabilization time is approximately two seconds. A bidirectional RS-232 interface with a second communications port offers connection to a printer for GLP/GMP compliant print-outs. The die-cast aluminum housing offers superior durability and chemical resistance. The sealed keypad, splashproof design, and removable stainless steel weighing pan offer additional safeguards in a messy lab environment. A lock down mounting point offers additional security from theft.
An oversized backlit LCD display provides clear visibility of weighing results. A three-door glass draft shield provides easy access inside the weighing chamber, while providing a safe environment for stable weighing. Digital filters compensate for vibration, drafts, or temperature changes.Cole-Parmer's precision analytical Symmetry balances are built with the most advanced weighing
technology available today, offering durability, and easy-to-use operation.
What's Included:
AC power adapter and draft shield
Specifications
Product Type
Analytical Balances
Auto shutoff
Yes
Calibration
Internal
Capacity (g)
220
Capacity (oz)
8
Housing dimensions
25.4 x 27.3 x 50.8 cm
Dimensions (" W)
10
Dimensions (" H)
10-3/4
Dimensions (" D)
20
Pan size (dia)
3-1/2" (8.9 cm)
Readability (g)
0.0001
Linearity (g)
±.0002
Repeatability (g)
±.0002
Weighing units
g, ct, oz, mg, dwt, grains
Stabilization time
2 to 4 seconds
Power (VAC)
115
Stability Indicator
Yes
Output
RS-232
Brand
Symmetry
Accessories
Qty
/
Price
Availability
Below Balance Hanger for Density Determination
EW-10000-37
at
$25.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Kensington
-type security lock and cable
EW-10000-47
at
$28.00 USD/EACH
In stock.
Add to Cart
ASTM Class 1 Mass with Weight Calibration Certificate, 100 g
EW-11954-20
at
$145.00 USD/EACH
In stock.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Symmetry Analytical Balance User's Manual (359 KB)
Cole-Parmer Symmetry Balances (973.33 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gravitational Effects on Balance
Weighing Unit Conversion Table
Common Balance Terms
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title