United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
11 >
EW-97621-10
Cole-Parmer UV Viewing Cabinet with 254nm and 365nm Wavelength Lamps, 115VAC
Click to enlarge
More Product Details
Specifications
Replacement parts
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-97621-10
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Lighting and Illumination
>
Ultraviolet Equipment
>
Ultraviolet Viewing Cabinets
>
Cole-Parmer Ultraviolet Viewing Cabinets
Share
|
Cole-Parmer UV Viewing Cabinet with 254nm and 365nm Wavelength Lamps, 115VAC
EW-97621-10
$1,935.00 USD/EACH
Usually ships in 15 days.
Qty
:
Add to Cart
Save to shopping list
Uniform UV illumination for optimal sample viewing
Soft rubber viewport provides a comfortable viewing area with adequate space for eyeglasses
Removable bottom panel allows cabinet to be placed over large samples or a transilluminator
Lightweight access curtain blocks out ambient light. Contrast control filter absorbs UV energy to protect users' eyes
High-performance viewing cabinet features high-intensity 15-watt UV tubes. Control switches, for selecting UV or white light, are conveniently located on the top of the cabinet. Model features two each of shortwave and longwave UV tubes mounted on one side of the cabinet.
Specifications
Product Type
Ultraviolet Viewing Cabinets
Watts
4 x 15
Wavelength
254/365 nm
Intensity
1200/950µW/cm
2
Dimensions
23.375 in W x 13 in H x 15.75 in D
Power
115VAC, 60Hz
Brand
Cole-Parmer
Manufacturer number
95-0504-04
Replacement parts
Qty
/
Price
Availability
365nm Wavelength Replacement Tube for Cole-Parmer UV Viewing Cabinets, 15-Watts
EW-97621-45
at $28.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
UV Equipment Terminology
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title