United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
32 >
SI-32460-44
Representative image only. Check individual details.
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Technical information
Virtual Catalog (PDFs)
More info
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
SI-32460-44
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17080-00
NIST-Traceable Calibration; Air/Gas Flowmeter
$235.00 USD/EACH/EACH
Usually ships in 10 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Flow, Level & Valves
>
Flowmeters
>
Variable Area Flowmeters
>
Cole-Parmer Valved Acrylic Flowmeters
>
Cole-Parmer Valved Acrylic Flowmeters for Bench or Panel Mount with Direct Metric-Reading Scales
Share
|
Cole-Parmer Valved Direct Reading Acrylic Flowmeter for Air Applications; Metric, 50-mm Scale, 0.4-5 LPM
SI-32460-44
$64.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Meter with valve provides flow control through a highly durable meter body
Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
Integrated precision valve allows precise manual flow across the full scale
The flexible design allows for panel or bench mounting
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately) which allows for mobility from bench to bench.
Note:
There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialists for quotes on acrylic meters with special requirements.
What's Included:
Flowmeter
Specifications
Product Type
Variable Area Flowmeter
Description
Variable Area, Direct Reading Flowmeter for Air
Max pressure (bar)
6.9
Max pressure (psi)
100
Accuracy
±5% full-scale
Material (fitting)
Chrome-Plated Brass
Material (Valve)
Brass
Material (o-ring)
Buna-N
Material (Housing)
Acrylic
Material (float)
316 Stainless Steel
Max operating temperature (º F)
150
Max operating temperature (º C)
65
Process connection
1/8" NPT(F)
Repeatability
±0.5% full-scale
Brand
Cole-Parmer
Dimensions
1"W x 4"H x 2-1/8"D
Manufacturer number
FR2A14BVBN-CP
Accessories
Qty
/
Price
Availability
Tripod base, mounts one flowmeter with 2" or 50-mm scale. Includes three leveling screws and spirit level.
EW-32462-50
at
$64.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Air/Gas Flowmeter
EW-17080-00
at $235.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Technical Resources
Mounting_Variable_Area_Flow_Meter.pdf
Virtual Catalog (PDFs)
13-14 Catalog
More info
Proper Mounting of your Variable Area Flow Meter.pdf
Related technical articles (Roll over for summary)
When to Choose Variable-Area Flow Meters
RotametersVariable Area Flowmeters Tech Info
Flowmeters- Why less is More
Show All...
Velocity-Profile Deviations Influence Flowmeter Performance
Selecting the Right FlowmeterPart 1
Selecting the Right FlowmeterPart 2
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title