United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-73160-40
Model 73160-00 pictured as an example only
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-73160-40
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Transfer Pump
>
Cole-Parmer Variable-Flow Chemical Transfer Pump
Share
|
Cole-Parmer Variable-Flow Chemical Transfer Peristaltic Tubing Pump
EW-73160-40
$287.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Variable-flow chemical transfer pump
Different wetted parts may be available upon request
Designed specifically for pulseless fluid transfer at variable flow rates. Pumps from 4-1/4 fluid ounces to 5/8 gallon (120 to 2,250 milliliters) per minute. Pumps fluids with a viscosity to 200 centipoises. Suction lift is 10 feet wet, 4 inches dry. May be used with fluid temperatures from –40 to 200°F (–40 to 93°C). Barbed inlet/outlet ports use any type of tubing with an inside diameter (nominal 3/16 inches).
Direct-drive engineering provides maximum motor power to the pump. Continuous-sweep variable control provides precise, seamless speed control. The chemical-resistant, wetted parts are Dupont Delrin®, 304 stainless steel, Viton, and Dupont PTFE®. Pump has a purge/prime switch for high-speed emptying/filling. It also reverses for ease in draining tubing. May be used with up to 120 feet of tubing for remote sampling. Pump comes ready to use with 115 VAC UL-rated wall power supply. Packaged in a chemical-resistant ABS plastic case. Size is 8 x 4-3/4 x 4-1/2 inches and weight is 3-1/4 pounds.
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title