United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
9 >
EW-79755-02
Cole-Parmer® Vacuum Pump Oil
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
MSDS
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-79755-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Pump Accessories
>
Vacuum/Compressor Pump Accessories
>
Oils and Fluids
>
Cole-Parmer Vacuum Pump Oil
Share
|
Cole-Parmer pump oil, 56 cSt, 1 gallon
EW-79755-02
$39.00 USD/GALLON
Quantity Discounts
Order
Discount
Price
[X]
4
10%
$35.10/GALLON
12
15%
$33.15/GALLON
24
20%
$31.20/GALLON
In stock.
Qty
:
Add to Cart
Save to shopping list
Cole-Parmer vacuum pump oil is formulated to ensure good performance and long pump life. CP 500 is a general-purpose high-viscosity oil. Use CP 19 in direct drive pumps to resist gum formation and oxidation. CP 20 exhibits a low vapor pressure and resists oxidation and thermal degradation—ideal for mechanical vacuum pumps. All types have a specific gravity of 0.86 g/mL.
Discounts: Save 10% on 12-23 quarts, 15% on 24-35 quarts and 20% on 36 or more quarts. Save 10% on 4-11 gallons, 15% on 12-23 gallons and 20% on 24 or more gallons.
Specifications
Product Type
CP 500
Vapor pressure
2x10
-4
torr at 25ºC
Viscosity
56 cSt
Brand
Cole-Parmer
Manufacturer number
CPGP GAL (INLAND 43)
Technical Resources
MSDS 1 (135 B)
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title