United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-08649-00
08649-10 image shown
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-08649-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17060-00
NIST-Traceable Calibration; Timer/Stopwatch
$55.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Timers
>
Digital Timers
>
Cole-Parmer Alarm Timers
>
Cole-Parmer Clock/Timers with Jumbo Display
Share
|
Cole-Parmer two-channel, jumbo display clock/timer
EW-08649-00
$26.00 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
5
10%
$23.40/EACH
10
20%
$20.80/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
This timer allows you to count up to or down from 100 hours with 1-second resolution. This timer features a 3/4" high display and multiple channels that can be used simultaneously.
Each channel has a distinctive alarm that sounds for 1 minute, pressing any button will turn off the alarm. Timer memory recalls previous time set. Features time-of-day in 12- or 24-hour formats.
What's Included:
magnetic clip, stand, and button cell battery.
Specifications
Timing capacity
100 hr
Product Type
Digital Multiple Channel Timers
Display
LCD, 3/4"H
Count direction
up/down
Clock
Yes
Alarm
1 min beep
Resolution
1 sec
Power
1.5 V button cell
No. of channels
two
Dimensions
2-3/4"W x 2-1/2"H x 1/2"D (7.0 x 6.4 x 1.3 cm)
Accuracy
±0.01%
Brand
Cole-Parmer
Manufacturer number
8913CP
Accessories
Qty
/
Price
Availability
Batteries, 1.5V button cells, 6/PK
EW-09377-16
at
$15.50 USD/PACKAGE of 6
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Timer/Stopwatch
EW-17060-00
at $55.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Instruction Manual
Timers Brochure(387 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Basics of Cartridge Filtration
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title