log in or register
Compact Ductless Air Cleaner, 350W, Black
EW-33703-39
$1,324.00 USD/EACH
Usually ships in 10 days.
Qty
  • Versatile system lets you customize to fit your application.
Capture and remove particles and volatile contaminants at their source. Compact air cleaner weighs only 30 lbs and fits virtually anywhere. Twin self-supporting extractor arms are made of durable polypropylene—they resist heat, chemicals, and abrasions. Ideal for two-station setups.
What's included: one 2-ply polyester ring-panel prefilter, one filter hold-down grid, one inlet grate, one inlet cover for two extractor arms, two extractor arms, two arm-hood attachments, and one arm-hole cap.
Complete your system with filters and accessories sold separately.
Product Type Fume hood extractor systems
Exterior Dimensions (" W) 12.5
Exterior Dimensions (" H) 12.5
Exterior Dimensions (" D) 19.5
Power (VAC) 120
Power (Hz) 60
Exhaust Volume (cfm) 350
Accessories
Qty/Price
Availability
at $169.00 USD/EACH
Usually ships in 14 days.
at $293.00 USD/EACH
Usually ships in 6 days.
at $210.00 USD/EACH
Usually ships in 5 days.
at $320.00 USD/EACH
Usually ships in 6 days.
at $455.00 USD/EACH
Usually ships in 7 days.
at $89.00 USD/PACKAGE of 4
Usually ships in 6 days.
at $45.00 USD/EACH
Usually ships in 8 days.
at $43.00 USD/EACH
Usually ships in 5 days.
at $108.00 USD/PACKAGE of 4
Usually ships in 10 days.
Dolly with casters
EW-33703-50
at $139.00 USD/EACH
Usually ships in 8 days.
Acrylic hood, 6" deep
EW-33703-52
at $166.00 USD/EACH
Usually ships in 5 days.
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title