Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Compact lab mixer

EW-50801-00
$1,030.00 USD/EACH

  • Use virtually anywhere—compact design and panel light make it perfect for enclosed workstations or fume hoods!
This mixer provides accurate and repeatable results every time. The brushless DC motor maintains power at all speeds and has automatic overload protection, so you can stir bigger batches longer. The belt driven transmission is smooth and quiet, and offers high-power efficiency. Stir up to 6.6 gallons (25 liters), and a maximum viscosity of 15,000 cps. The splash-proof sealed housing keeps liquids and moisture out—good for aggressive environments. The through-shaft design makes adjustments easy and decreases downtime due to impeller changes. Adjustable chuck accommodates up to 3/8" diameter impeller shaft.

Mixer features an integrated lighting system for illuminating solutions so they can be clearly seen. The four-button control (power on/off, speed increase/decrease, light on/off) makes this mixer extremely easy to use. Order stand, clamp, and mixing impellers separately.

Compact lab mixer includes:
– stainless steel chuck (accepts up to 3/8" dia shafts) and key
– 5"L x 1/2" dia mounting rod
– external power supply
– 6-ft power cord from power supply with three-prong plug

Product Type Mixer
Dual range No
Min speed (rpm) 40
Max speed (rpm) 2000
Motor (hp) 0.07
Mounting type universal clamp
Programmable No
Remote control No
Reversible No
Shaft dia (Max) 0.375 in
Max torque (in-oz) 100 (70 N-cm)
Power (VAC) 120/230
Power (Hz) 50/60
Overload protection Yes
Torque (Max) 100
Computer interface No
Explosion-proof No
Recorder compatible No
Hazardous duty No
Continuous duty Yes
Constant speed No
Folding Impeller No
double helix mixing No
Illuminating Yes
Dual-shaft No
Through-shaft Yes
CE Compliance Yes
Display Digital
Brand Caframo

Accessories
Qty/Price
Availability

at $128.00 USD/EACH

Usually ships in 10 days.

at $112.00 USD/EACH

In stock.

at $202.00 USD/EACH

Usually ships in 8 days or sooner.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 13 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title