United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
EW-44204-00
Compact ultra-low freezer, benchtop, 115 VAC, 60 Hz
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-44204-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Cooling Equipment
>
Freezers
>
ScienTemp Compact Ultra-Low Freezers
Share
|
Compact ultra-low freezer, benchtop, 115 VAC, 60 Hz
EW-44204-00
$4,395.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
An ultra-low freezer that can sit on the benchtop!
Key locking door provides additional sample security
These compact ultra-low freezers are ideal for clinical trial and small sample storage. They fit easily on the benchtop or undercounter without taking up a lot of space. Freezers have a temperature range of –50 to –86°C and provide 1.0 cubic feet of storage space. Microprocessor control provides temperature accuracy of ±1°C and features an audible alarm and diagnostic capabilities.
Cabinets include CFC-free, foamed-in-place insulation that conserves energy and protects samples. Both benchtop and undercounter models have the display located at the top of the unit for easy viewing along with a top-mounted compressor. Chart recorders may be ordered separately. Freezers will accommodate up to four storage racks.
What's Included:
an 8-ft cord with 3-prong plug on 115 VAC models only.
Specifications
Product Type
Benchtop Freezers
Capacity (cu ft)
1
Capacity (Liters)
28
Dimensions (interior)
12-1/4"W x 11"H x 13"D
Dimensions (overall)
20"W x 28-1/2"H x 22"D
Min Temp - Freezer (º C)
-86
Min Temp - Freezer (º F)
-122
Temp accuracy
±1°C
Power (Hz)
60
Power (Breaker)
10 A
Cabinet construction
heavy-gauge steel with powder coat finish
Display
4-digit LED
Brand
ScienTemp
Manufacturer number
86-01A
Power (VAC)
115
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
LABORATORY EQUIPMENT- Getting Cryogenics Down Cold
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title