Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Constant-torque brushless mixer; 40 to 6000 rpm, 115 VAC

EW-50800-20
$1,505.00 USD/EACH

Usually ships in 12 days or sooner.
Qty

  • Chuck guard encloses spinning chuck for added safety
  • Set speed maintained as fluid viscosity changes
  • Includes tare function to set torque reading at zero
The unique, two-speed transmission delivers constant performance under changing condistions. Mixer features a sparkless, brushless DC motor that provides a long, maintenance-free service life. The totally enclosed non-ventilated motor is rated for continuous duty. Overload protection automatically shuts off the mixer if maximum torque is exceeded—an LED light signals overload conditions. The through-shaft design lets you easily alter shaft length and propeller depth. Mixers have an aluminum alloy housing.
What's included: calibration certificate, stainless steel chuck and key (accepts up to 3/8'' dia shafts), chuck guard, 5''L x 1/2'' dia mounting rod, and 6-ft cord with three-prong plug.
Product Type Mixer
Dual range Yes
Min speed (rpm) 40
Max speed (rpm) 6000
Motor (hp) 0.2
Motor type TENV
Mounting type universal clamp
Programmable No
Remote control No
Reversible No
Shaft dia (Max) 0.375 in
Max torque (in-oz) Low-speed range: 120
High-speed range: 48
Power (VAC) 115
Overload protection Yes
Torque (Max) 240
Computer interface No
Explosion-proof No
Recorder compatible No
Hazardous duty No
Continuous duty Yes
Constant speed Yes
Folding Impeller No
double helix mixing No
Illuminating No
Dual-shaft No
Through-shaft Yes
Display Digital
Brand Caframo

Accessories
Qty/Price
Availability

at $128.00 USD/EACH

Usually ships in 10 days.

at $112.00 USD/EACH

In stock.
Mixer safety stand
EW-50001-93

at $226.00 USD/EACH

In stock.

at $195.00 USD/EACH

In stock.

at $209.00 USD/EACH

Call 1-888-358-4705 for availability.

at $249.00 USD/EACH

Usually ships in 7 days.
Calibrations
Qty/Price
Availability

at $115.00 USD/EACH
Usually ships in 13 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title