Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Contact plate, TSA with Lecithin and Tween 80, 10/pk
Item# EW-14201-20
$25.75 USD/BAG
Are you a reseller or buying in bulk? Call us today.
Usually ships in 4 days.
Qty
  • Cultivate microorganisms from environmental surfaces
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
These high-quality, economical plates feature a specified grid molded into the bottom of the plate for easy colony counting.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Contact Plates
Description TSA with Lecithin and Tween 80, for the cultivation of microorganisms from environmental surfaces
Qty/pk 10
Media Name Media A-C
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:
      Can I extend the expiration date by performing growth promotion on a lot of contact plate, TSA with Lecithin and Tween 80?
      Asked on 2/2/2013 by None
      1 answer
      • STAFF EXPERT
        A:
        We would not recommend using the plates past expiration date.
        Answered on 2/5/2013 by James from Technical Support
    • Q:
      What are the storage requirements for these plates?
      Asked on 12/10/2010 by Anonymous
      1 answer
      • STAFF EXPERT
        A:
        Store these plates in a refrigerator within a temp range of 2-8 Celsius.
        Answered on 12/13/2010 by Mike from Tech Support
    Displaying questions 1-2
    Do you have a question about this product? 
    Product Reviews
    REVIEW SNAPSHOT®
    by PowerReviews
    Contact plate, TSA with Lecithin and Tween 80
    4.0
    (based on 1 review)
    Reviewed by 1 customer
    Displaying review 1
    Back to top
    4.0
    MORE BANG FOR THE BUCK
    By Chris
    from Dillon,S.C.
    Verified Buyer
    Pros
    • value
    • Works Great
    Cons
      Best Uses
      • CHECKING FOR BACTERIA
      • CHECKING FOR MOLD
      • SANITATION PLATING
      Comments about Contact plate, TSA with Lecithin and Tween 80:
      use this product in our hatchery to test bacteria and mold levels. Was using from another company who charge way to much. SO I switch and these things have done the job.
      Displaying review 1
      Back to top
      Customers Who Bought This Item Also Bought