Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Contec Prosat Presaturated cleanroom wipes, 300/pack

EW-33675-00
$95.00 USD/PACKAGE of 300

  • Ideal for general-purpose cleaning in Class 100 environments
Prosat wipes measure 9" x 11" and are presaturated in a solvent blend of 30% DI water and 70% isopropyl alcohol; solvents are filtered to a 0.2 micron level. Low-lint polypropylene wipes won’t scratch delicate surfaces. Wipes come in a convenient pack of 30 that can be easily ransported and used gain. Wipes are sold in cases of 10 packs of 30 wipes/pk.


Note: Not available for sale to the state of California


This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Wipes
Compliance Class 100
Brand Contec
Manufacturer number PS-911, PROSAT
Model PS-911, PROSAT





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title