United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Controller for Masterflex Liqui Sense emergency cut off system 115 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Build a complete system
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-77096-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Tubing and Pump Accessories
>
Liqui-Sense Emergency Cut-Off System
>
Liqui-Sense Controller
Share
|
Controller for Masterflex Liqui-Sense emergency cut-off system, 115 VAC.
EW-77096-00
$585.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Monitor pumps and fluid systems for leaks, flow, and fluid level24 hours a day
Shuts down pump or process automaticallyeliminating downtime, system damage, and fluid loss
Customize to your specific application needs using dual input and output channels
Controller will shut off the main pump and switch on a backup pump when a sensor detects a problem.
Audible and visible alarms alert you when problems are detected; a remote jack lets you connect an external alarm.
Select from three types of sensors: liquid detector pad senses leaks; out-of-liquid sensor detects air in tubing; float-switch level sensors detect level changes in tanks and vessels.
Controller has connections for two sensors each with their own sensitivity adjustment (controller provides 12 VDC, 20 mA per channel for powering sensors). Monitor two points in a single system or two independent systems.
Purchase optional environmental monitor (model 08330-20 separately) to have your system call your phone or pager in the event of problems.
Model 77096-00 has 115 VAC main and backup pump connections with U.S. standard plug receptacles; 10 A, 50/60 Hz. Includes a 6-ft (1.8 m) line cord with U.S. standard plug.
The complete Liqui-Sense
®
system consists of the controller, a sensor, and an environmental monitor (optional).Optional extension cables 77095-03 and 77095-04 allow you to position the sensors up to 15 feet (4.5 m) from the controller.
Specifications
Product Type
Accessory
Control type
Digital Variable Speed
Brand
Masterflex
Model
77096-00
Manufacturer number
77096-00
Accessories
Qty
/
Price
Availability
Environmental monitor, 4 inputs, for use with Masterflex Liqui-Sense system.
EW-08330-20
at
$499.00 USD/EACH
Usually ships in 5 days.
Add to Cart
Interface cable for monitor 08330-20; length, 20-ft
EW-23000-70
at
$19.50 USD/EACH
Usually ships in 14 days.
Add to Cart
Optional extension cable, RJ12; 10 ft (3 m).
EW-77095-03
at
$55.00 USD/EACH
In stock.
Add to Cart
Optional extension cable, RJ12; 15 ft (4.5 m).
EW-77095-04
at
$58.00 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Build a complete system
1
.
Environmental Monitor
Details
2
.
Liqui-Sense Controller
Details
3
.
Liquid Sensors
Details
Technical Resources
Instruction Manual (429 KB)
Instruction Manual (607 KB)
Instruction Manual (620 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Liqui-Sense Emergency Cutoff
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Show All...
Peristaltic Tubing Pumps in Dispensing Applications
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title