Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

CoorsTek Porcelain Unglazed Rose Crucible, 75 mL; 6/Pk

CoorsTek  |   Mfr# 60166   |   Item# EW-17809-00

$75.00 USD/PACKAGE of 6

Are you a reseller or buying in bulk? Call us today.


    Plain crucible without glaze. Ideal for general drying applications. Autoclavable.
    Capacity (mL) 75
    Top OD (mm) 50
    Material Porcelain
    Manufacturer number 60166
    Brand CoorsTek
    Qty/pk 6





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:

      Does this crucible come with a lid?
      Asked on 10/29/2012 by Tony

      1 answer

      • STAFF EXPERT

        A:

        No, this does not come with a lid. At this time CoorsTek does not make a lid for this size and unglazed.

        Answered on 10/30/2012 by Jen from Technical Support
    • Q:

      I would like to know if they can withstand temp. up to 2800F or not.
      Asked on 2/10/2011 by Ben

      1 answer

      • STAFF EXPERT

        A:

        No, this crucible will not withstand temperatures up to 2800 F. This crucible will withstand temperatures up to 2552 degrees F (1400 degrees C).

        Answered on 2/14/2011 by Jaime from Technical Support
    Displaying questions 1-2

    Do you have a question about this product? 

    Product Reviews