Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Corning 96-Well Black Solid Plates, Nontreated

EW-01728-66
$395.00 USD/CASE of 100

Usually ships in 15 days.
Qty

  • Tested on a lot-to-lot basis for opacity and low background fluorescence/luminescence—assures minimal well-to-well crosstalk
  • Choose Black Polystyrene plates for minimizing background fluorescence and backlight scatter.
  • Well volume is 360 µL.
Product Type Microplates
Surface treatment Untreated
Color Black
Well shape Flat
Well volume (mL) 0.36
Well format 96 well
Brand Corning
Manufacturer number 3915

Accessories
Qty/Price
Availability
Acetate sealing tape
EW-01730-00

at $56.00 USD/CASE of 100

Call 1-888-358-4705 for availability.

at $214.00 USD/CASE of 100

Call 1-888-358-4705 for availability.

at $80.00 USD/CASE of 50

Call 1-888-358-4705 for availability.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title