Corning cell culture flask, 25 cm2, canted neck, plug seal cap, sterile
EW-01936-00
$905.00 USD/CASE of 500
Are you a reseller or buying in bulk? Call us today.
Usually ships in 10 days.
Qty
  • Available in a variety of sizes, designs and cap styles to meet your needs
  • Virgin polystyrene provides excellent optical quality
  • Gamma radiated; certified nonpyrogenic and nontoxic
  • Barcoded model available
Flasks have individually printed lot numbers for product traceability. Feature molded-in graduations. Anti-tip skirt provides increased stability on 25-, 75-, and 150-cm2 flasks.
Capacity (mL) 30
Neck type Canted
Brand Corning
Manufacturer number 430168
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title