United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
Representative photo only
Davis Instruments Mounting Tripod for Weather Stations 5 8 ft tall
Click to enlarge
More Product Details
Specifications
Related products
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-86403-70
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Weather Products
>
Weather Stations
>
Davis Instruments Professional Weather Station Mounting Tripod
Share
|
Davis Instruments Mounting Tripod for Weather Stations, 5.8 ft tall
EW-86403-70
$75.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Brackets at the base of each leg tilt to mount on your roof or uneven terrain
Galvanized steel construction
Use a mounting tripod for your Davis Vantage Pro2 or Vantage Vue to make installation even easier. Includes two 3-foot (0.92-meter) poles, which may be used separately or together to make a single 5.8-foot (1.77-meter) pole.
Available for sale in the United States and Canada only.
What's Included:
two 3-foot (0.92-meter) poles and mounting hardward
Specifications
Product Type
Weather Station accessories
Model
7716
Manufacturer number
7716
Related products
Qty
/
Price
Availability
Davis Instruments Vantage Vue Wireless Station, includes console and sensor suite.
EW-30001-97
at $395.00 USD/EACH
In stock.
Add to Cart
Davis Instruments Vantage Pro2
Wireless Weather Station
EW-86403-03
at $595.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual (860 KB)
Instruction Manual.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title