United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
6 >
Representative photo only
Davis Instruments WeatherLink Software with data logging feature for Vantage Pro 2 USB
Click to enlarge
More Product Details
Specifications
Related products
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-86403-54
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Weather Products
>
Weather Stations
>
Davis Instruments Vantage Pro2 Weather Station Accessories
Share
|
Davis Instruments WeatherLink Software (with data logging feature) for Vantage Pro 2, USB
EW-86403-54
$165.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
WeatherLink
®
software with data logger allows you to graph, analyze and share your data with weather watchers around the world
Add WeatherLink to your Vantage Vue or Vantage Pro2. The data logger fits neatly into the weather station console or Weather Envoy, storing weather data even when it's not connected to your PC.
Add WeatherLink to any Vantage station or Weather Envoy and store weather data even when it's not connected to a computer. Later, download the data and use the software for detailed analysis and graphing. Our popular WeatherLink with standard data logger is ideal for most purposes, while other models add special features designed for specific user groups.
Expanded Reporting
• More Weather Data.
View weather information not available on the console, including heating degree days, cooling degree days, and fuel-oil demand.
• Optional Sensor Reporting.
With optional UV and solar radiation sensors, get details on sunburn risk and solar energy.
• Moon Phase.
With Vantage Pro2 and Vantage Vue, you can see the phase of the moon right on the console. But now even Perception and Weather Wizard III owners can view the phases of the moon using WeatherLink.
Sharing Your Weather Data
• Create Your Own Reports.
Data can be exported to most popular spreadsheets, databases, and word processing programs.
• Weather Website.
Create a weather website or post weather conditions to a LAN. Add graphs, text lines, or scrolling ticker tapes, and upload other files (such as webcam images) along with weather data.
• Weather World 'Round.
Register your URL at davisnet.com and we'll add it to our growing list of Davis stations around the world.
• Citizen Weather Observer Program.
Submit your data to the National Weather Service to help develop improved weather prediction models.
• The GLOBE Program.
Participate with other schools in an international weather-related program for students from elementary through high school.
What's included:
software on CD, data logger and cable to connect the data logger to your computer. Add Extra User License Kits for additional computers.
Available for sale in the United States and Canada only.
Specifications
Product Type
Davis Vantage Pro2
Brand
Davis Instruments
Model
6510USB
Manufacturer number
6510USB
Related products
Qty
/
Price
Availability
Davis Instruments Vantage Pro2
Wireless Weather Station
EW-86403-03
at $595.00 USD/EACH
In stock.
Add to Cart
Davis Instruments Vantage Pro2
Plus Wireless Weather Station with UV and solar radiation sensors
EW-86403-13
at $995.00 USD/EACH
In stock.
Add to Cart
Technical Resources
86403-54_Setup_Installation.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Monitoring Changing Conditions: Weather Stations
Temperature FAQs
Temperature Instrument Ranges
Show All...
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title