Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Decon Labs Bacdown Hand Soap, 1 L Bottle

Bacdown  |   Mfr# 7001  |   Item# EW-86115-06

$31.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


    Gentle handsoap combines soothing, coconut-based surfactants with skin-kind Triclosan to provide a handsoap that may be used repeatedly with minimal skin drying. It is recommended for applications where repeated hand scrubbing is necessary.
    Product Type Hand Cleaner
    Format Pump
    Volume (Liters) 1
    Packaging 1 Liter
    Model 7001
    Qty/ea 1
    Manufacturer number 7001
    Brand Bacdown





    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      Do you have MSDS for this item

      Thanks
      Asked on 1/28/2010 by Anonymous

      1 answer

      • STAFF EXPERT

        A:

        Please contact us directly (phone or email) for electronic file requests. AnswerBox does not currently have the functionality to send files.

        Answered on 1/29/2010 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews

    REVIEW SNAPSHOT®

    by PowerReviews
    Bacdown<reg> Hand Soap, 1 L
     
    4.5

    (based on 2 reviews)

    Reviewed by 2 customers

    Sort by

    Displaying reviews 1-2

    Back to top

     
    4.0

    GREAT PRODUCT

    By MITCH

    from NEW JERSEY

    Verified Buyer

    Pros

    • Cleans Well
    • HELPFUL FOR DRY SKIN

    Cons

    • COSTLY FOR HOME USE

    Best Uses

    • WE USE AT HOME

    Comments about Bacdown<reg> Hand Soap, 1 L:

    WE PURCHASED THIS ITEM FOR MY DAUGHTER WHO HAD A SLIGHT CASE OF ECZAMA AND WHO WASHES HER HANDS OFTEN. AFTER USING THIS PRODUCT WE SAW A BIG DIFFERENCE IN HER HAND'S APPERANCE. THE ONLY THING IS THAT THIS IS QUITE A COSTLY ITEM ARE THERE ANY DISCOUNT PLANS

     
    5.0

    Great Stuff

    By Student Ivan

    from WPI,MA

    About Me Student

    Verified Buyer

    Pros

    • Cleans Quickly
    • Cleans Well
    • Sanitizes

    Cons

    • No fruity smells

    Best Uses

    • Home use
    • In The Field
    • In The Lab
    • Research

    Comments about Bacdown<reg> Hand Soap, 1 L:

    wash my hands at least every 15 minutes and my hands never get dry or cracked with this product! It's amazing! No complaints! Maybe fruity smells for the future?

    Displaying reviews 1-2

    Back to top

    Customers Who Bought This Item Also Bought