United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
Defenders Stainless Steel Receptacle 7 gallon
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-47580-08
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Handling and Storage
>
Document and Waste Containers
>
Defenders Stainless Steel Receptacles
Share
|
Defenders Stainless Steel Receptacle, 7 gallon
EW-47580-08
$303.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
All models are UL and FM listed
Receptacles are non-magnetic and will not carry a charge, and will never rust or corrode. Defenders
®
step cans comply with OBRA Regulations and OSHA standards for the safe containment of infectious waste. All models are fire safe and self-extinguishing and California State Fire Marshal listed (except model C-47580-06). Square models are standard with poly bag retainer bands that can be used in conjunction with galvanized steel and rigid plastic liners, and also have a piano hinged lid restraint mechanism to protect walls and equipment. Round models come complete with vinyl base rings that protect floors and reduce noise. Replacement plastic liners are also available; order at right.
Specifications
Product Type
Waste Bins
Dimensions
12 in x 12 in x 17 in
Dimensions (" W)
12
Dimensions (" H)
17
Dimensions (" L)
12
Material
Stainless Steel
Capacity (gal.)
7
Brand
Defenders
Manufacturer number
ST7SSPL
Model
ST7SSPL
Accessories
Qty
/
Price
Availability
Repalcement liner for SS Receptacle 47580-08
EW-47580-22
at
$40.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title