Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Desco 09041, Adjustable Metal Speidel Wristband without Cord
Desco  |   Mfr# 09041  |   Item# EW-26809-00
$35.50 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 7 days.
Qty
  • Protect delicate electronics from static damage
Wristbands and straps have 4-mm stainless steel snap studs. Purchase with or without a coiled cord; cords are 6-ft long and include a 1-MΩ resistor. Cleanroom compatible and safe for microelectronics applications. One size fits all. UL-listed.
Speidel® Twist-O-Flex® Wristbands feature a 300-series stainless steel interior expansion band. Flexible design won't pinch or grab hair.
Elastic Adjustable Wristbands feature a wide, soft elastic band and 300-series stainless steel used for all metal parts.
Product Type Wrist Straps
Qty/ea 1
Brand Desco
Model 09041
Manufacturer number 09041
Accessories
Qty/Price
Availability
at $18.50 USD/EACH
Usually ships in 8 days.
at $27.75 USD/EACH
Usually ships in 8 days.
at $130.50 USD/EACH
Usually ships in 5 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456