log in or register
Dewar Flask with Handles, 4300mL
EW-03771-13
$435.00 USD/EACH
  • This flask incorporates the convenience of a coated handle
  • Increased portability and pouring efficiency
  • Dewar lip is extended to facilitate pouring
  • Vented polyethylene lid helps to minimize evaporation
These Dewars feature an exterior metal housing with a boroscilicate glass interior. There are no pads between walls to eliminate heat transfer potential. These are ideal for temporary storage or for transfer applications. Dewar flasks come with lids.
Product Type Dewars
Volume (Liters) 4.3
Dimensions (" OD) 7
Dimensions (" ID) 4.6875
Depth 12
Accessories
Qty/Price
Availability
Lid F/03771-13
EW-03771-92
at $8.00 USD/EACH
Usually ships in 5 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title