United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
Representative photo only
Digi Sense 1 Zone benchtop dual display temperature control type J input 120V 15 amps
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-36225-61
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17000-10
NIST-Traceable Calibration; Thermocouple Meter (meter only)
$115.00 USD/EACH/EACH
Usually ships in 7 days.
EW-17002-10
NIST-Traceable Calibration; Thermocouple Thermometer System (meter + probe)
$160.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Temperature, Humidity, Moisture and Weather Products
>
Temperature Products
>
Temperature Controllers
>
Controllers-Benchtop
>
Digi-Sense Benchtop PID Temperature Controllers
Share
|
Digi-Sense 1-Zone benchtop dual display temperature control, type J input, 120V, 15 amps
EW-36225-61
$625.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Perfect for basic temperature control applications
Models available for type J, K, or T thermocouples or RTD probes
These temperature controllers display both the setpoint and process temperature and features modern control technology that produces greater temperature stability. The Auto-tune feature minimizes setpoint overshoot and features the ability to learn your process producing greater stability. Changing your setpoint is easily done with the interface keys. The controller can also perform ramp rate operations to allow the user to slowly raise the process temperature. The controller also features an audible alarm output, temperature selectable and one panel mounted receptacle that accept a three-prong plug. Select from models for types J, K, or T thermocouples with miniconnectors, or 100 ohm platinum RTD probes. This unit comes with a lattice support bracket, which is ideal for mounting to maximize space.
What's Included:
6’ detachable power cord and a lattice support bracket
Specifications
Product Type
Temperature control, benchtop
Control input (thermocouple)
Yes
Control output 1
SSR
Control output
SSR
Min temperature (º C)
-18
Max temperature (º C)
65
Min temperature (º F)
0
Max temperature (º F)
149
Dimensions (" W)
8
Dimensions (" H)
3-3/8
Dimensions (" D)
6
Display
LED
DIN size
1/16"
Resolution
1°, 1.0° or 1.00°
Accuracy
0.1% of span, ±1.0°F/°C
Brand
Digi-Sense
Manufacturer number
104A 3622561
Power (VAC)
120
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Thermocouple Meter (meter only)
EW-17000-10
at $115.00 USD/EACH
Usually ships in 7 days.
Add to Cart
NIST-Traceable Calibration; Thermocouple Thermometer System (meter + probe)
EW-17002-10
at $160.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Temperature Control: An Overview and Product Selection Guide
Temperature Controller Features
Electron zap turns flimsy plastic into sturdy shrink wrap
Show All...
Basics of Cartridge Filtration
Bitter Brew
Temperature FAQs
Temperature Instrument Ranges
Getting a Handle on Temperature
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title