Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Digi-Sense Calibrated 2-in-1 Waterproof Infrared Thermometer
Item# EW-35625-41
$101.00 USD/EACH
$80.80 USD/EACH
Are you a reseller or buying in bulk? Call us today.
  • A thermometer for all applications
  • Go/no-go LEDs provide quick check of HACCP food zones
  • IP65 rated waterproof housing
Ultra thin 2-in-1 waterproof contact/noncontact thermometer features a HACCP food safety LED warning. LED’s rapidly indicate if food temperature is within a safe or unsafe range. Green light indicate safe temperatures below 40°F (4°C) and above 140°F (60°C). Red light indicates the HACCP danger zone of 40 to 140°F (4 to 60°C). The built-in stainless steel type-K thermocouple probe swings out for internal temperature checks, then folds in for storage. IR mode provides quick, noncontact surface measurements. Bright white LED pointer illuminates the target measurement area and is ideal for close working distances of 2 to 12”. Product also features adjustable emissivity, Celsius or Fahrenheit selectable, min/max, auto data hold, lock
Product Type Infrared Thermometer
Distance to target size ratio 2.5:1
Max temperature (º C) 250
Min temperature (º C) -55
Min temperature (º F) -67
Max temperature (º F) 482
Accuracy (º C) IR: ±0.1°C from -33 to 0°C, ±1°C from 0 to 65°C, ±1.5% of reading from 65 to 300°C, Probe: ±0.5°C from -9 to 100°C else ±0.9% of reading
Accuracy (º F) IR: ±0.1°F from -27 to 32°F, ±1°F from 32 to 149°F, ±1.5% of reading from 149 to 572°F, Probe: ±0.5°F from 15 to 212°F else ±0.9% of reading
Emissivity Adjustable
Sighting type LED (non-laser)
Power two AAA batteries (included)
Display 4-digit LCD, 0.75 in W x 1 in H
Low battery indicator Yes
Qty/ea 1
Manufacturer number 7199DS
CE Compliance Yes
Model 7199DS
Brand Digi-Sense
Accessories
Qty/Price
Availability
at $24.75 $19.80 USD/PACKAGE of 12
In stock.
at $7.25 USD/BOX of 100
In stock.
Calibrations
Qty/Price
Availability
at $136.00 $105.00 USD/EACH
Usually ships in 5 days.
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456