Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

Digi-Sense Oven Thermometer, 0/200°C, 405mm Length, 125mm Immersion

Item# EW-08006-57

$32.25 USD/EACH
$27.41 USD/EACH

Are you a reseller or buying in bulk? Call us today.

  • Replace mercury thermometers for a specific application with the same accuracy without the any hazards
  • Non-mercury liquid
  • Swell
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PTFE coated for added safety which may cause a slight change in accuracy up to 1 division.

Oven thermometers - designed for high temperature applications and feature a round glass swell to hold the thermometer in place
Product Type Glass Thermometer
Min temperature (º C) 0
Max temperature (º C) 200
Division (º C) 1
Length (mm) 405
Material Glass
Accuracy ±2°C; ±2.5°C above 100°C
Special features Swell in glass to hold thermometer in place
Fill Red Spirit
Model 608030730
Qty/ea 1
Brand Digi-Sense


at $166.00 USD/EACH
Usually ships in 11 days.

Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    1 Question | 1 Answer
    Displaying question 1
    • Q:

      Can this thermometer be used with oven to replace the mercury filled one which is place on a thermomether holder on top?
      Asked on 9/14/2011 by Sylvia Enid from Puerto Rico

      1 answer

      • Staff Reviewer


        It may be able to replace it. This thermometer has a 125mm immersion depth. So, the tip will need to be in the oven to the specified line in order to provide and accurate reading. If the thermometer tip is not in far enough, or in too far you will not see an accurate temperature reading.

        Answered on 9/15/2011 by Jen from Technical Support
    Displaying question 1

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought