Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Digi-Sense Thermowell, 316 Stainless Steel, 18" Length, 3/4" Connection
Item# EW-90434-04
$76.00 USD/EACH
$68.40 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 3 days.
Qty
  • Designed for easy replacement of your industrial probe without shutting the operation down.
To match your probe to a thermowell, first measure the distance from the probe’s tip to the top of the thread (typical thread length is 0.5" for 1/2" NPT fittings). Next, match this length to the probe depth of the thermowell (“A” dimension). Other thermowell sizes and materials are available—call our Application Specialists for more information.
Limited-Space Thermowell when you need a shorter insertion depth.
Standard Thermowell for general-purpose applications.
Heavy-Duty Thermowell for higher fluid velocity ratings.
Product Type Thermowell
Max temperature (º C) 871
Min temperature (º F) -32
Max temperature (º F) 1600
Min temperature (º C) 0
Model 6000-H-3/4-16.5-0-0
Qty/ea 1
Brand Digi-Sense
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456