Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Digi-Sense Type K Fine-gauge, Bare-wire thermocouple probes; 0.005" diameter, 5/pack

EW-08419-01
$26.00 USD/PACKAGE of 5

Quantity Discounts
Qty:
Price:
6
$23.40
11
$22.10
25+
$20.80
    Fine-gauge, bare-wire probes with 0.005" and 0.010" diameters provide quick temperature measurements (time constant typically less than 0.3 seconds). Larger probes with 0.020", and 0.032" diameters ensure long-term durability but have a slower response time (time constant typically 1.5 seconds or less). Probes feature beaded junctions and 12" leads. The negative lead is slightly shorter than the positive lead for easy identification. Probes come in packs of five.
    Min temperature (º F) -328
    Max temperature (º F) 2282
    Max temperature (º C) 1250
    Probe length (in.) 12
    Diameter (mm) 0.1
    Diameter (inches) 0.005
    Temp range -328 to 2282°F (-200 to 1250°C)
    Probe type K
    Min temperature (º C) -200
    Time constant 0.10 sec
    Junction beaded
    Manufacturer number 120301D
    Brand Digi-Sense
    Model 120301D
    Product Type Thermocouple Probe

    Calibrations
    Qty/Price
    Availability

    at $130.00 USD/EACH
    Usually ships in 7 days.

    at $160.00 USD/EACH
    Usually ships in 6 days.




    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  • 13-14 Spanish Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought