United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-25750-60
Digital Density / Specific Gravity Meter
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-25750-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Density/Specific Gravity
>
Digital Density/Specific Gravity Meter
Share
|
Digital Density / Specific Gravity Meter
EW-25750-60
$3,775.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Compact design and wide grip for easy one-hand operationperfect for on-site inspections
This density meter is easy to grip and control even when you're wearing thick work gloves. The narrow 1" (2.5 cm) depth lets you maneuver in tight spaces. Meter features a pipette-style pump for good sample flow controlno bellows or bulb to rupture and leak. To draw in a sample, simply press the lever at the top of the meter with your thumb. View density, specific gravity, % concentration, or sample temperature on the large LCD. Press lever again to discharge sample.
Store up to 1024 readings with a push of a button. Access permanently stored concentration tables including % Brix, % alcohol, proof, °Baume, °Plato, API gravity, specific gravity at reference temperature, and % H
2
SO
4
. Durable meter housing is sealed against pump spills. Infrared interface lets you send stored readings to your PC for later analysisorder infrared to USB adaptor or optional infrared printer separately. Case is included.
Specifications
Specific gravity range
0 to 3 g/cm
3
or SGU
Specific gravity resolution
0.0001 g/cm
3
or SGU
Sample volume
2 mL
Temp range
32 to 104°F (0 to 40°C)
Temp resolution
0.1°F (0.1°C)
Output
Infrared interface
Dimensions
5-1/2" x 5-3/8" x 1" (14.0 x 13.6 x 2.5 cm)
Power
Two 1.5 V AA batteries (included)
Accessories
Qty
/
Price
Availability
Batteries, 1.5 V, AA, 4/pack
EW-09376-01
at
$7.50 USD/PACKAGE of 4
In stock.
Add to Cart
Infrared to USB Adapter
EW-25750-61
at
$69.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Infrared Printer for Density / Specific Gravity Meter
EW-25750-62
at
$391.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Electron zap turns flimsy plastic into sturdy shrink wrap
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title