United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
Digital Humidity Controller with Temperature
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-37700-03
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Environmental Chambers
>
Chamber Accessories
>
Humidity Controllers
Share
|
Digital Humidity Controller with Temperature
EW-37700-03
$2,520.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Maintain humidity levels with high accuracy controllers
Continuously control humidity levels to your desired set points
Controllers accurately measure and control the humidity level in your humidity or environmental chambers. Choose model 37700-02 to control both humidifier and dehumidifier, or 37700-03 to control humidity and display temperature.
Controllers accurately measure and control the humidity level in your humidity or environmental chambers. Model 37700-03 controls humidity and displays temperature.
Model 37700-03 activates with 0.1% deviation from the set point. It also has an RTD temperature sensor with a range of -40 to 140°F (-40 to 60°C). Controllers are housed in ABS plastic and are user-selectable for 115 or 230 VAC operation.
What's Included:
NIST-traceable calibration report supplied by the manufacturer, humidity sensor with a 5-ft (1.5 m) cable, a U.S. standard powered receptacle rated at 4 A or less, and a 7 1⁄2-ft (2.3 m) power cord with three-prong plug.
Specifications
Display
Dual LEDs
Temp accuracy
±0.36°F (0.2°C)
Humidity range
0 to 100%
Humidity resolution
0.1%
Humidity accuracy
±1.5%
Dimensions (overall)
7 1/4"W x 2 1/2"H x 9" D
Output
0 to 1 VDC, RS-232
Set point tracking
±0.2%
Temp range
-40 to 185°F (-40 to 85°C)
Temperature resolution
0.1°F/°C
Power
90 to 240 VAC, 50/60 Hz
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title