United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
5 >
Representative photo only
Digital Manometer with range of 0 to 5 psi
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68603-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-31
NIST-Traceable Calibration; Manometers
$140.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Digital and Analog Manometers
>
Digital Manometer with Backlight Display
Share
|
Digital Manometer with range of 0 to 5 psi
EW-68603-00
$259.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Read gauge and differential pressure in 11 units of measure!
Ideal for measuring low differential pressure in clean rooms, test and balance, HVAC, pneumatic systems, and computer peripherals
Large backlight LCD, maximum, minimum, hold, and auto power off functions
Built-in RS-232 port downloads information to your PC
Order cable and software separately from Accessories below
What's Included:
connection hose, 9 V battery and hard shell carrying case.
Specifications
Product Type
Manometers
Media compatibility
non-corrosive air and gases
Accuracy
±0.3% full-scale
Unit of Measurement
psi, " H
2
O, kPa, ft H
2
O, " Hg, cm H
2
O, mbar, bar, mm Hg, oz inch
2
, kg cm
2
Resolution
0.003 psi
Output
RS-232
Range
0 to 5 psi
Process connection
3/16" barb
Power
one 9 V battery (included)
Display
4-digit, 1/2"H LCD with backlight
Accessories
Qty
/
Price
Availability
AC Power Adapter; 115 VAC
EW-68603-20
at
$17.50 USD/EACH
Usually ships in 7 days or sooner.
Add to Cart
RS-232 cable, cable rs232-interface
EW-68603-22
at
$39.00 USD/EACH
In stock.
Add to Cart
Software, software manometer data
EW-68603-24
at
$94.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Manometers
EW-17090-31
at $140.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Anemometers
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Show All...
Pressure Unit Conversion Constants
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title