Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Digitrate Electronic Burettes; 30 mL capacity

EW-25201-05
$952.00 USD/EACH

Usually ships in 8 days.
Qty

  • Glass barrel lets you visually inspect reagent
  • Simple user calibration and factory reset
Digital readout displays meniscus-free total volume readings—using these electronic burettes is so easy. To move from one titration to the next, simply note your final reading, then press the zero switch to continue to the next titration. Low gearing enables dispensing in single increments of 0.01 mL with accuracy better than ±0.2%. The readout is not affected during refilling—dispense up to 99.99 mL without interruption.
Comes with a lifetime battery that powers more than 60,000 titrations. Optional biological filter is sold separately. Order 1400-mL or 2500-mL bottles.
What’s included: dispense nozzle protection sleeve; lithium battery; threaded adapters for 33-, 38-, and 45-mm bottle necks; calibration test certificate.
Product Type Electronic Burettes
Tolerance ±0.2%
Capacity (mL) 30
Brand Digitrate
Manufacturer number 182-000
Model 182-000

Calibrations
Qty/Price
Availability

at $66.00 USD/EACH
Usually ships in 9 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title