log in or register
Digitrate PRO, 50Ml, Electronic Burette
EW-25205-10
$906.00 USD/EACH
  • Delivers accurate, precise, and convenient bottle-top titration
  • Includes calibration certificate with individual serial number
Features continuous display of volume dispensed with zero reset for multiple titrations. New safety pedestal valve allows the fluid path to be turned off manually to prevent accidental release. Accuracy is achieved within ± 0.2%, precision better than 0.1%, resolution 0.04mL. Head can be rotated 360° for operator safety and comfort. Unit will automatically switch off without losing previous titrated volume. Parts can be dissassembled for sterilization or autoclaving at 121°C. Unit is powered by a 3.6v lithium battery; allowing a minimum life of 60,000 three-minute titrations. Includes 33mm, 38mm, and 45mm adapters.
Product Type Digital Burettes
Capacity (mL) 50
Brand Digitrate
Manufacturer number 182-026
Model 182-026
Accessories
Qty/Price
Availability
at $12.00 USD/EACH
Usually ships in 8 days.
at $12.00 USD/EACH
Usually ships in 8 days.
at $12.00 USD/EACH
Usually ships in 8 days.
Digitrate Pro 45 mm
EW-25201-63
at $14.00 USD/EACH
Usually ships in 8 days.
at $124.00 USD/EACH
Usually ships in 8 days.
at $182.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability
at $66.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title