United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
8 >
EW-36150-00
Dilution bottle, Butterfield's phosphate buffer, 99 mL fill capacity
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-36150-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Microbiology
>
Dilution Bottles
>
Prefilled Dilution Bottles
Share
|
Dilution bottle, Butterfield's phosphate buffer, 99 mL fill capacity
EW-36150-00
$94.00 USD/PACKAGE
Usually ships in 4 days.
Qty
:
Add to Cart
Save to shopping list
The dilution solution saves time and money
Wide 45-mm opening accommodates large samples
Flip-top lid allows for easy one-handed opening
Prefilled dilution vials are designed for the food, dairy, pharmaceutical, and water industries. Eliminate the time and money spent on preparing your own dilution vials. Final pH for all solutions is 7.2 ±0.2 at 25°C. The 145-mL polypropylene vial features a flip-top lid for easy one-handed opening. The peptone water vials are gamma irradiated.
Butterfield's Phosphate Buffer
is used as a diluent for the preparation of dilutions in plate count and other laboratory processes. It is recommended as a general diluent in laboratory procedures by the Federal Drug administration and in the Bacteriological Analytical Manual. This product is described in the Standard Methods for the Examination of Water and Wastewater for use in water testing. Contains deionized water and monopotassium phosphate.
Specifications
Product Type
Dilution bottles
Application
Foods, dairy products, water, wastewater
Fill capacity
99 mL
Material
Polypropylene
Capacity (Liters)
0.145
Capacity (mL)
145
Qty/pk
50
Brand
Hardy Diagnostics
Technical Resources
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title