dummy
EW-07055-52
Usually ships in 14 days.
Qty:
- Four-vane rotary design is almost maintenance free
- Nonsparking, nonelectrical operation—safe and reliable
What's Included:
Motors come with an exhaust muffler. Install a compressed-air filter in-line to remove oil, water, and solid contaminants—order connecting nipples and fliters separately.
| Product Type |
NEMA air motors |
| hp |
1 |
| Speed range |
350 to 3000 rpm |
| Shaft dia |
5/8" |
| Shaft centerline |
3-1/4" |
| Max cfm |
30 |
| Frame size |
56C |
| Dimensions |
5-7/8"L x 6-1/2"W x 6-1/2"H |
at $110.00 USD/EACH
In stock.
at $12.50 USD/PACKAGE of 6
Call 1-888-358-4705 for availability.
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection