Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Home  > 
Discontinued Items >
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.
Disposable Antistatic PP Micro Scoops, 3-7 mg, Green; 100/Pk
Item# EW-06277-54
$20.75 USD/PACKAGE
Quantity Discounts
Qty: 6 30 120+
Price: $18.68 $17.64 $16.60
Discontinued
Qty
    Designed to transfer powders in small quantities, micro scoops improve your accuracy, precision, and efficiency. Scoop container doubles as an easy-to-use dispenser to reduce cross contamination. Available in economical, anti-static polypropylene or durable stainless steel. Autoclavable.
    Product Type Scoops
    Overall length 5.5 in
    Capacity (mL) .02
    Material Polypropylene
    Technical Resources
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Do you have questions about this product?
    Get answers from real customers and in-house experts with AnswerBox.
    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:
      Is this product one size or four sizes?
      Asked on 12/17/2013 by BigPasta from Tallahassee, FL
      1 answer
      • STAFF EXPERT
        A:
        Each scoop is the same size, and depending on the density of the powder, the mass contained will weigh anywhere from 3-7 mg.
        Answered on 12/18/2013 by Dan from Technical Support
    • Q:
      The description states that these are also available in stainless steel. Can you give me the part number for these micro scoops in stainles steel (and pricing)?
      Asked on 7/13/2013 by Gregg from The Ohio State University
      1 answer
      • STAFF EXPERT
        A:
        The SS version would be available in the 6-8 mg size - please see part number 00310DE
        Answered on 7/15/2013 by Jen from Technical Support
    Displaying questions 1-2
    Do you have a question about this product? 
    Product Reviews
    REVIEW SNAPSHOT®
    by PowerReviews
    Micro Scoop PP 3-7 mg
    4.0
    (based on 1 review)
    Reviewed by 1 customer
    Displaying review 1
    Back to top
    4.0
    Good Product for the Price
    By analyticalchemguy
    from Reading, PA
    Verified Buyer
    Pros
    • Cheap per item
    • Consistent measurements
    • Disposible
    Cons
    • Sometimes sticks
    Best Uses
      Comments about Micro Scoop PP 3-7 mg:
      Works great for a quick and consistent measurement for small amounts of powder. The cheap price per unit is nice so you do not have to worry about saving to clean to reuse. Simply use once and discard. The only drawback that I found is that despite the product being anti-static sometimes powder gets caught in the bottom of the scoop dish and is difficult to remove. You may have to measure twice in some instances.
      Displaying review 1
      Back to top
      Customers Who Bought This Item Also Bought