United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
EW-08022-60
Image is representative of item.
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-08022-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Clamps, Supports, Laboratory Jacks
>
Troemner Clamps and Supports
>
Utility Clamps
>
Research-Grade Double Burette Utility Clamps
Share
|
Double Buret Clamp/Stand
EW-08022-60
$214.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Available in stainless steel or nickel plated zinc
An additional holder must be purchased in order to attach clamps to lab-frames, support stands or other apparatus
Hold one or two burettes up to 100mL. The one-handed, scissor-like mechanism provides quick and easy adjustment. Clamp is available separately or together with stand.
Hold one or two burettes up to 100 mL. The one-handed, scissor-like mechanism provides quick and easy adjustment. Includes support base, rod and clamps.
Specifications
Product Type
Burette Clamps
Description
Double buret holder & rod, with base. Buret clamp holds (2) burets from micro to 100mL
Dimensions (" L)
22.76
Material (Base)
Porcelain
Material (Support rod)
Aluminum
Material (Clamp)
Nickel-plated Zinc
Brand
Troemner
Accessories
Qty
/
Price
Availability
Thumbscrew Knob
EW-0804795
at
$38.00 USD/EACH
Usually ships in 11 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title