log in or register
Drager Accuro Pump Kit
EW-86514-00
$568.00 USD/EACH
  • Easy, one-handed operation
  • Wider diameter tubes with well-spaced graduation marks are easier to read
The accuro® gas detector pump is a manually operated bellows pump that draws a calibrated 100 mL sample through the short-term Dräger gas detection tubes. Provides fast, accurate, reliable results in just seconds.
Product Type Air/Gas Analysis Kits
Brand Drager
Manufacturer number 4053473
Accessories
Qty/Price
Availability
at $87.00 USD/BOX
Usually ships in 6 days.
at $81.00 USD/BOX
Usually ships in 5 days.
at $87.00 USD/BOX
Usually ships in 7 days.
at $81.00 USD/BOX
In stock.
at $92.00 USD/BOX
In stock.
at $108.00 USD/BOX
Usually ships in 14 days.
at $87.00 USD/BOX
Usually ships in 2 days.
at $87.00 USD/BOX
Usually ships in 6 days.
at $73.00 USD/BOX
In stock.
at $75.00 USD/BOX
Usually ships in 3 days.
at $76.00 USD/BOX
Usually ships in 5 days.
at $77.00 USD/BOX
Call 1-888-358-4705 for availability.
at $81.00 USD/BOX
Usually ships in 7 days.
at $139.00 USD/BOX
Usually ships in 14 days.
at $75.00 USD/BOX
Usually ships in 6 days.
at $95.00 USD/BOX
Usually ships in 6 days.
at $95.00 USD/BOX
Usually ships in 10 days.
at $114.00 USD/BOX
Usually ships in 7 days or sooner.
at $95.00 USD/BOX
Usually ships in 4 days.
at $82.00 USD/BOX
In stock.
at $72.00 USD/BOX
In stock.
at $76.00 USD/BOX
In stock.
at $115.00 USD/BOX
Usually ships in 8 days.
at $76.00 USD/BOX
Usually ships in 4 days.
at $174.00 USD/BOX
Usually ships in 8 days.
at $87.00 USD/BOX
In stock.
at $87.00 USD/BOX
In stock.
at $77.00 USD/BOX
In stock.
at $81.00 USD/BOX
Usually ships in 6 days.
at $95.00 USD/BOX
Usually ships in 6 days.
at $110.00 USD/BOX
Usually ships in 14 days.
at $88.00 USD/BOX
Usually ships in 14 days.
at $108.00 USD/BOX
Usually ships in 11 days.
at $87.00 USD/BOX
Call 1-888-358-4705 for availability.
at $81.00 USD/BOX
In stock.
at $96.00 USD/BOX
Usually ships in 7 days.
at $96.00 USD/BOX
In stock.
Drager Tubes, Alcohol
EW-86514-61
at $97.00 USD/BOX
In stock.
at $87.00 USD/BOX
Usually ships in 6 days.
at $88.00 USD/BOX
Usually ships in 7 days.
at $75.00 USD/BOX
In stock.
at $76.00 USD/BOX
Usually ships in 4 days.
at $81.00 USD/BOX
Usually ships in 12 days.
Technical Resources
  • Instruction Manual.pdf
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title